missing translation for 'onlineSavingsMsg'
Learn More

Restek™ High-capacity and ECD Vent Traps

Catalog No. 06710915
Encompass
Change view
Click to view available options
Type:
ECD Vent Trap
High Capacity Split Vent Trap
Packaging Quantity:
1 Each
5/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Quantity
06710915 High Capacity Split Vent Trap 1 Each
06711822 ECD Vent Trap 1 Each
06717795 High Capacity Split Vent Trap 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06710915 Supplier Restek™ Supplier No. RK 20698

Includes connecting lines and mounting kit.

  • High-Capacity Split Vent Trap reduces the release of hazardous materials from the capillary split vent into the lab
  • ECD Vent Trap reduces the release of hazardous materials from the ECD vent into the lab
  • Fittings: 1/8 in.
  • Dimensions: 15.2 x 2.5cm (6 x 1 in.)

Specifications

Connection Type 0.12 in. Fitting
Type High Capacity Split Vent Trap
Includes Connecting lines and mounting kit
Packaging Quantity 1 Each
Length (English) 6 in.
Length (Metric) 15.2 cm
Diameter (English) Outer 1 in.
Diameter (Metric) Outer 2.5 cm