missing translation for 'onlineSavingsMsg'
Learn More

Cytiva RESOURCE™ S IEX Columns

Catalog No. p-3676541
Change view
Click to view available options
Volume (Metric):
1 mL
6 mL
Quantity:
1 x 1 mL
1 x 6 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
45000199 1 mL 1 x 1 mL
45000201 6 mL 1 x 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000199 Supplier Cytiva Supplier No. 17117801
Only null left

RESOURCE prepacked columns for fast separation. Resource S IEX Columns are prepacked with Source 15S, a strong cation exchanger for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.

  • Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25mg/mL and stability in organic solvents and pH extremes
  • Resource columns with 1mL bed volume have separation times of 3 minutes or less at 9.6mL/min. and approximately 20 min. using a peristaltic pump at 1mL/min.

Specifications

Description RESOURCE S
Kit Contents Connectors for high- and low-pressure chromatography equipment and instructions
Column Material PEEK
Matrix Polystyrene/divinylbenzene
Flow Rate >10 mL/min.
For Use With (Application) RESOURCE S columns are prepacked with SOURCE 15S, a strong cation exchanger for high-resolution polishing purification of proteins, at high flow rates.
Ligand Type Sulphonate group
Content And Storage 4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol
Diameter (Metric) Inner Column 6.4 mm
Volume (Metric) 1 mL
Particle Size 15 μm
pH 2 to 13
Quantity 1 x 1 mL
Show More Show Less