missing translation for 'onlineSavingsMsg'
Learn More

Cytiva RESOURCE™ Q IEX Columns

Catalog No. 45000198
Change view
Click to view available options
Volume (Metric):
1 mL
6 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
45000198 1 mL
45000200 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000198 Supplier Cytiva Supplier No. 17117701

Use for intermediate lab-scale purification and large-scale polishing of biomolecules Cytiva Life Sciences™ RESOURCE™ Q IEX Columns are strong anion exchanger columns.

  • Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25mg/mL and stability in organic solvents and pH extremes
  • Resource columns with 1mL bed volume have separation times of 3 minutes or less at 9.6mL/min. and approximately 20 min. using a peristaltic pump at 1mL/min.

Specifications

Description Dynamic Capacity: 40-54 mg BSA/ml
Kit Contents Connectors for high- and low-pressure chromatography equipment and instructions
Column Material PEEK
Matrix Polystyrene/divinylbenzene
Flow Rate >10 mL/min.
For Use With (Application) RESOURCE Q columns are prepacked with SOURCE 15Q, a strong anion exchanger for high resolution polishing purification of proteins at high flow rates.
Ligand Type Quaternary ammonium
Content And Storage 4°C to 30°C in 20% Ethanol
Diameter (Metric) Inner Column 6.4 mm
Volume (Metric) 1 mL
Particle Size 15 μm
pH 2 to 12
Quantity 1 x 1 mL
Show More Show Less