missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Resistin Monoclonal Antibody (K94029_2B12), Invitrogen™
Mouse Monoclonal Antibody
$1,740.58 - $2,926.56
Specifications
| Antigen | Resistin |
|---|---|
| Clone | K94029_2B12 |
| Concentration | 0.62 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: SSKTLCSMEE AINERIQEVA GSLIFRAISS IGLECQSVTS RGDLATCPRG FAVTGCTCGS ACGSWDVRAE TTCHCQCAGM DWTGARCCRV QP
Resistin Monoclonal antibody specifically detects Resistin in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| Resistin | |
| 0.62 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Adipose tissue specific secretory factor; adipose tissue-specific secretory factor; Adipose-specific cysteine-rich secreted protein A12-alpha; ADSF; C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 1; C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich protein precursor 1; c/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; cysteine-r; Cysteine-rich secreted protein A12-alpha-like 2; cysteine-rich secreted protein FIZZ3; dominant inhibitory adipocyte-specific secretory factor; dri; FIZZ3; found in inflammatory zone 3; HXCP1; MGC126603; MGC126609; Resistin; resistin delta2; RETN; RETN1; RSN; RSTN; UNQ407/PRO1199; XCP1; Xcp4 | |
| Recombinant human RETN (immunogen range is 17-108). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| RETN |
| K94029_2B12 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9HD89 | |
| 56729 | |
| IgG1 | |
| Protein A | |
| Antibody |