missing translation for 'onlineSavingsMsg'
Learn More

Residual Solvents Mixture - Class IIA Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017011
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017011 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017011 Supplier Merck Emd Millipore Supplier No. PHR10643X1.2ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Residual Solvents Mixture - Class IIA are certified reference materials (CRMs) for the analysis of residual solvents. The residual solvents can be found as mixtures, as single-analyte standards, as well as neat CRMs. These CRMs are secondary pharmaceutical reference standards and are traceable and qualified against the corresponding Pharmacopeial standards.

Specifications

CAS 110-82-7
Flash Point 87°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Type Certified Reference Material
Quantity 3 x 1.2 mL
Grade Certified Reference Material