missing translation for 'onlineSavingsMsg'
Learn More

Residual Solvents Class 1 PT, MilliporeSigma™ Supelco™

Catalog No. 111001683
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001683 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001683 Supplier Merck Emd Millipore Supplier No. PEP0181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Specifications

Packaging Ampule
CAS 71-43-2
Flash Point 87°C - closed cup
Type Proficiency Testing Material
Quantity 1 g