missing translation for 'onlineSavingsMsg'
Learn More

Reserpine 98.0+%, TCI America™

Catalog No. R00071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00071G 1 g
R00075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00071G Supplier TCI America Supplier No. R00071G

Chemical Identifiers

CAS 50-55-5
Molecular Formula C33H40N2O9
Molecular Weight (g/mol) 608.688
MDL Number MFCD00005091
InChI Key QEVHRUUCFGRFIF-MDEJGZGSSA-N
Synonym reserpine, serpalan, serpasil, serpivite, apoplon, deserpine, elserpine, hiserpia, hypersil, raunervil
PubChem CID 5770
ChEBI CHEBI:28487
IUPAC Name methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyl)oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
SMILES COC1C(CC2CN3CCC4=C(C3CC2C1C(=O)OC)NC5=C4C=CC(=C5)OC)OC(=O)C6=CC(=C(C(=C6)OC)OC)OC

Specifications

Formula Weight 608.69
Percent Purity ≥98.0% (HPLC,T)
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Reserpine
EINECSNumber (9)-1522
RTECSNumber ZG0350000
TSCA Yes