accessibility menu, dialog, popup

UserName

CAS RN 50-55-5

IUPAC Name: methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyloxy)-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate
Synonyms: reserpine recipin V-serp hydroserp rauserpin-alk cam-ap-es demi-regroton klimanosid serpivite serpate
Molecular Weight (g/mol): 608.69
Molecular Formula: C33H40N2O9
InChi Key: QEVHRUUCFGRFIF-MDEJGZGSSA-N
SMILES: CO[C@H]1[C@@H](C[C@@H]2CN3CCC4=C(NC5=CC(OC)=CC=C45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)C1=CC(OC)=C(OC)C(OC)=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)