missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reserpine Standard for LC-MS, 5 mg/L in Water: Isopropyl Alcohol (1:1), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 50-55-5 |
| Flash Point | 22°C |
| Linear Formula | C33H40N2O9 |
| MDL Number | MFCD00005091 |
| Melting Point | 265°C |
| Synonym | Reserpine; (3 beta, 16 beta, 17 alpha, 18 beta, 20 alpha)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester |
| Type | Analytical Standard |
| Quantity | 4.5 mL |
| Formula Weight | 608.68 |
| Grade | Analytical Standard |
| Show More |