missing translation for 'onlineSavingsMsg'
Learn More

Reserpine, 99%, Thermo Scientific Chemicals

Catalog No. p-7058274
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0350603 1 g
AAL0350606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0350603 Supplier Thermo Scientific Chemicals Supplier No. L0350603

Inhibits vesicular uptake of catecholamines and serotoninReserpine is an antipsychotic and antihypertensive drug used to treat the hypertension, strokes, heart attacks and kidney problems. It is used to treat the symptoms of dyskinesia in patients who are affected by huntington’s disease. It is widely used as a reference material in the mass spectroscopy technique and also used as a sedative for horses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Incompatible with reducing agents and oxidizing agents.

Chemical Identifiers

CAS 50-55-5
Molecular Formula C33H40N2O9
Molecular Weight (g/mol) 608.688
MDL Number MFCD00005091
InChI Key QEVHRUUCFGRFIF-MDEJGZGSSA-N
Synonym reserpine, serpalan, serpasil, serpivite, apoplon, deserpine, elserpine, hiserpia, hypersil, raunervil
PubChem CID 5770
ChEBI CHEBI:28487
IUPAC Name methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyl)oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
SMILES COC1C(CC2CN3CCC4=C(C3CC2C1C(=O)OC)NC5=C4C=CC(=C5)OC)OC(=O)C6=CC(=C(C(=C6)OC)OC)OC

Specifications

Melting Point ∼265°C
Beilstein 102014
Merck Index 14,8145
Solubility Information Soluble in water.
Formula Weight 608.69
Percent Purity 99%
Quantity 1 g
Chemical Name or Material Reserpine
Hazard Category H302+H312+H332-H350-H360D
Hazard Statement GHS H Statement
H301-H360-H351
Toxic if swallowed.
May damage fertility or the unborn child.
Suspected of causing cancer.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 200-047-9
RTECSNumber ZG0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only