missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes

Catalog No. 05414265
Change view
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Color:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
05414265 0.25 to 2.5 mL Red
13690033 0.5 to 5 mL Violet
13690029 10 to 100 μL Yellow
13690025 0.1 to 2.5 μL Dark Gray
13690032 100 to 1000 μL Blue
13690030 20 to 200 μL Yellow
13690028 2 to 20 μL Yellow
13690034 1 to 10 mL Turquoise
13690027 2 to 20 μL Light Gray
13690026 0.5 to 10 μL Medium Gray
13690031 30 to 300 μL Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 05414265 Supplier Eppendorf™ Supplier No. 3123000144

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Adapt for specific liquids or other external conditions with these ultra-light fully autoclavable pipettes. Pipettes feature a spring loaded tip cone, a secondary adjustment option and an improved volume display.

  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejectors low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display
  • Calibration display and adjustment opening for specific liquids and volumes
  • Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
  • Quick lock makes lower part particularly easy to remove and service (not available for 5 and 10 mL pipettes)
  • Spring loaded tip cone promotes tight fit to tip (not available for 5 and 10 mL pipettes)
  • Has ability to be autoclaved without disassembling

Applications

  • Pipetting liquids
  • Mixing liquids
  • Filling of plates, gels, and reaction vessels
  • Phase extraction and removal of supernatants

Specifications

Battery Type None
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Red
Model Eppendorf™ Research™ Plus™
Number of Channels 1
Product Line Research Plus
Type Single-Channel Adjustable Volume Pipettes
Volume (Metric) 0.25 to 2.5 mL
Display Digital
Finger Style Ergonomic
Includes epT.I.P.S.™ Sample Bag with 10 Pipette Tips
Show More Show Less