missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Research™ plus, Mechanical Single-Channel Pipettes PROMO

Catalog No. 13690026PM
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13690026PM 0.5 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13690026PM Supplier Eppendorf™ Supplier No. 2231001212
Get Up to 50% Off Eppendorf Equipment & Microtubes
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Upgrade your pipetting workflow with the perfect precision pairing. The Eppendorf Research™ plus Pipette and Repeater™ M4 Bundle combines ergonomic accuracy with high-efficiency repetitive dispensing – everything your lab needs to work faster, safer, and more consistently.

Eppendorf Research™ plus pipettes: single channel (10 μL,100 μL,200 μL,1000 μL)

  • Reliable and Ergonomic: Ideal for everyday pipetting with a lightweight design, low operating forces, and minimal strain
  • Autoclavable and Durable: Fully autoclavable for safety and long-term use, with high resistance to chemical and mechanical stress
  • Repeater™ M4 - (Combitips™ Advanced sold separately, 1 μL to 10 mL)
  • Aspirate once and dispense up to 100 times with high accuracy and efficiency - ideal for repetitive tube filling, plate filling, and reagent aliquoting
  • Positive displacement technology ensures reliable dispensing of viscous, volatile, dense, or hazardous liquids, while auto-recognized Combitips™ advanced and ViscoTip™ tips improve safety and ease of use

Specifications

Accuracy ±8%, ±2.5%, ±1.5%, ±1%
Compatible Tips 20 μL Pipette tips, 40 mm L
Autoclavable Autoclavable
Color Medium Gray
Disposable Reusable
For Use With Pipette Tips
Number of Channels 1
Product Line Research Plus
Sterility Non-sterile
Type Adjustable Single Channel Pipette
Volume (Metric) 0.5 to 10 μL
Display Digital
Finger Style Ergonomic
For Use With (Equipment) 20 μL Pipette Tip
Includes epT.I.P.S.™ Box 2.0 with 96 Pipette Tips
Increments 0.5, 1, 5, 10 μL
Voltage 0/1 V
Fit for Purpose Certified Quality
Show More Show Less