missing translation for 'onlineSavingsMsg'
Learn More

Resazurin Sodium Salt 85.0+%, TCI America™

Catalog No. p-7135754
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R02031G 1 g
R02035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R02031G Supplier TCI America Supplier No. R02031G
Only null left

Chemical Identifiers

CAS 62758-13-8
Molecular Formula C12H6NNaO4
Molecular Weight (g/mol) 251.173
MDL Number MFCD00005036
InChI Key IVGPGQSSDLDOLH-UHFFFAOYSA-M
Synonym resazurin sodium salt, resazurin, sodium salt, 7-hydroxy-3h-phenoxazin-3-one-10-oxide sodium salt, sodium 3-oxo-3h-phenoxazin-7-olate 10-oxide, sodium 7-oxophenoxazin-10-ium-3,10-bis olate, sodium 10-oxido-7-oxophenoxazin-10-ium-3-olate, sodium; 7-oxo-10-oxy-7h-phenoxazin-3-olate, sodium 10-oxo-10h-10??-phenoxazin-10-ylium-3,7-bis olate, resazurin sodium salt 1g
PubChem CID 112939
IUPAC Name sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate
SMILES C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+]

Specifications

Color Black-Green
Quantity 1 g
Formula Weight 251.17
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Resazurin Sodium Salt