missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Resazurin, sodium salt, pure, high purity biological stain
CAS: 62758-13-8 | C12H6NNaO4 | 251.173 g/mol
Chemical Identifiers
| CAS | 62758-13-8 |
|---|---|
| Molecular Formula | C12H6NNaO4 |
| Molecular Weight (g/mol) | 251.173 |
| MDL Number | MFCD00005036 |
| InChI Key | IVGPGQSSDLDOLH-UHFFFAOYSA-M |
| Synonym | 7-Hydroxy-3H-phenoxazin-3-one 10-oxide, sodium salt |
| PubChem CID | 112939 |
| IUPAC Name | sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate |
| SMILES | C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC189900010
|
Thermo Scientific Chemicals
189900010 |
1 g | Glass bottle |
N/A
|
|
||||
|
AC189900050
|
Thermo Scientific Chemicals
189900050 |
5 g | Glass bottle |
N/A
|
|
||||
|
AC189900250
|
Thermo Scientific Chemicals
189900250 |
25 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 62758-13-8 | |
| 251.173 | |
| IVGPGQSSDLDOLH-UHFFFAOYSA-M | |
| 112939 | |
| C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+] |
| C12H6NNaO4 | |
| MFCD00005036 | |
| 7-Hydroxy-3H-phenoxazin-3-one 10-oxide, sodium salt | |
| sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate |
Specifications
| 62758-13-8 | |
| C12H6NNaO4 | |
| 27, 128 | |
| 7-Hydroxy-3H-phenoxazin-3-one 10-oxide, sodium salt | |
| C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+] | |
| sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate | |
| 112939 | |
| Pure | |
| Black to Green | |
| Resazurin, sodium salt |
| 75% min. | |
| MFCD00005036 | |
| 14, 8141 | |
| IVGPGQSSDLDOLH-UHFFFAOYSA-M | |
| Authentic | |
| 251.173 | |
| 251.17 | |
| Complies | |
| Powder |
Safety and Handling
Warning
RUO – Research Use Only