missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Resazurin, sodium salt, pure, high purity biological stain

Catalog No. ac189900010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189900010 1 g Glass bottle
AC189900050 5 g Glass bottle
AC189900250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC189900010 Supplier Thermo Scientific Chemicals Supplier No. 189900010
Only null left

CAS: 62758-13-8 | C12H6NNaO4 | 251.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62758-13-8
Molecular Formula C12H6NNaO4
Molecular Weight (g/mol) 251.173
MDL Number MFCD00005036
InChI Key IVGPGQSSDLDOLH-UHFFFAOYSA-M
Synonym 7-Hydroxy-3H-phenoxazin-3-one 10-oxide, sodium salt
PubChem CID 112939
IUPAC Name sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate
SMILES C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+]

Specifications

Dye Content 75% min.
Beilstein 27, 128
Merck Index 14, 8141
Infrared Spectrum Authentic
Formula Weight 251.17
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Black to Green
Quantity 1 g
Physical Form Powder
Chemical Name or Material Resazurin, sodium salt
Show More Show Less
Hazard Category Warning

RUO – Research Use Only