missing translation for 'onlineSavingsMsg'
Learn More

REPS1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566010
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501566010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501566010 Supplier Bethyl Laboratories Inc Supplier No. A303769A
Only null left

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

May coordinate the cellular actions of activated EGF receptors and Ral-GTPases By similarity. Mungall A.J., Nature 425:805-811(2003). Mural R.J., Submitted (SEP-2005) to the EMBL/GenBank/DDBJ databases. Kimura K., Genome Res. 16:55-65(2006).

Specifications

Antigen REPS1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene REPS1
Gene Accession No. Q96D71
Gene Alias Cell migration-inducing gene 2 protein, Raft-linking protein, RFTN1, KIAA0084, ORF Names: MIG2
Gene Symbols REPS1
Host Species Rabbit
Immunogen Between 650 and 700
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85021
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less