Learn More
Representative Protist Set
All four classes of protist are represented.
Supplier:  Customer Channels Group (Ccg) S10036
Catalog No. S10036
Only null left
Explore available promotions
Description
Description
These specimen slides represent the common protozoa.
Includes:
Sarcodina
  • Amoeba proteus, w.m.
  • Pelornyxa carolinensis, w.m. of giant multinucleate amoeba
  • Entamoeba histolytic, trophozoites, smear; parasite causing amoebic dysentery in humans
  • Entamoeba histolytica, cysts, smear; each cyst contains 1-8 nuclei
  • Foraminifera, shells of various species, w.m.
Mastigophora
  • Trypanosoma gambiense, blood smear; parasite causing African sleeping sickness in man
  • Euglena
Ciliata
  • Ciliates of cow stomach, w.m.; several species of extremely complex ciliates which live as commensals in stomach of cow
  • Didimium, w.m. preys exclusively on Paramecium
  • Paramecium caudatum, Fission, w.m.
  • Stentor, w.m.; very large trumpet-shaped ciliate with beaded nucleus
  • Vorticella, w.m.; bell-shape, stalked ciliate
Sporozoa
  • Hemogregarina, smear of frog blood, in which many red cells have been invaded by this parasite
  • Monocystis, in section of earthworm seminal vesicle
Specifications
Specifications
Representative Protozoan
15/Pk.
Product Suggestions
Product Suggestions