Learn More
Corning™ PYREX™ Replacemet Parts for 6949M Series Microchemistry Kits
Catalog No. 11850106
Change view
Click to view available options
Product Type:
Conical Reaction Vial
Catalog No. Product Type
11850106 Conical Reaction Vial
1 options
Catalog No. 11850106 Supplier Corning™ Supplier No. C99001
Get Up to $2,500 in Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with PYREX™ Microchemistry Kits
This is a replacement part for the 6949M series of PYREX™ microchemistry kits.
Specifications
For Use With (Equipment) 6949M Series PYREX™ MicrochemistryKits
Includes Polyethylene case
Material Glass
Length (Metric) 44 mm
Capacity (Metric) 1 mL
Diameter (Metric) Outer 13.5 mm
Sterility Non-sterile
Product Type Conical Reaction Vial