missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Replacement Tube for Kuderna-Danish Apparatus

Catalog No. 0777071
Change view
Click to view available options
Capacity (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
0777071 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0777071 Supplier Corning™ Supplier No. 215710TO

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Replacement tube for use with Kuderna-Danish apparatus

These PYREX replacement tubes fit the Kuderna Danish Apparatus and have an outer 19/22 Standard Taper joint. The tube graduations go from 0 to 2 mL in 0.1 mL increments and from 3 mL on in 1.0 mL increments.

Specifications

Product Type Replacement Tube
For Use With (Equipment) Kuderna-Danish Apparatus
Capacity (Metric) 10 mL
Capacity (English) 0.3 oz.
Graduations 0 to 2 mL/0.1 mL, 3 mL/1 mL
Standard Taper Joint Top 19/22