missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Traceable™ Replacement Printing Paper
Description
- For use in Traceable™ Printing Thermohygrometers
Specifications
Specifications
| Material | Paper |
| Ruled | No |
| Width (English) Page | 1.5 in. |
| Color | White |
| For Use With (Equipment) | Traceable™ Printing Thermohygrometers |
| Product Type | Printer Paper |
| Quantity | 2 Rolls |