Learn More
BTX™ Replacement Pins for Plate Handler
Catalog No. BTX450469
Change view
Click to view available options
Quantity:
100 Pk.
25 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTX450469 100 Pk.
BTX450468 25 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX450469 Supplier BTX™ Supplier No. 450469
Replacement Pins designed to work with BTX™ Plate Handlers.
  • Replacement Pins for BTX™ Plate Handlers
Specifications
For Use With (Application) High Throughput electroporation or transfection
For Use With (Equipment) HT 100, HT 200
Material Gold-plated
Product Line BTX Electroporation
Product Type Electroporation Spare Pin
Description Replacement Pins for Plate Handler, Pkg. of 100
Electrode Shape Pin
Includes 100 x Replacement Pins
Quantity 100 Pk.
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Exponential Decay Wave, Square Wave
High-throughput Compatibility Yes
Show More Show Less