Learn More
SGE™ Replacement Needles for 5μL Syringes
Made of Stainless steel.
Supplier:  SGE™ 036110
 View more versions of this product
Catalog No. SG-036110
Only null left
Alert:
Made of Stainless steel
Description
Description
Made of Stainless steel
Specifications
Specifications
26 G
0.018 in.
0.47 mm
Replacement Needle
8 mm O.D. Barrel 5μL Microvolume Syringes
0.0043 in.
0.11 mm
1.9 in.
Bevel
Product Suggestions
Product Suggestions