Learn More
Welch™ Replacement Element for Vacuum Pumps
Accessory for Welch vaccum pumps
Supplier:  Welch™ 1420E01
 View more versions of this product
Catalog No. 0110415A
Only null left
Description
Description
  • For acid neutralization trap and oil mist elinimator
  • Changes from white to bluish-transparent when spent
Specifications
Specifications
Model 1402N, 1376N vacuum pump
Acid Neutralization Trap
Product Suggestions
Product Suggestions