Learn More
Welch™ Replacement Element for Vacuum Pumps
Accessory for Welch vaccum pumps
$410.80 - $521.59
Description
- For acid neutralization trap and oil mist elinimator
- Changes from white to bluish-transparent when spent