Learn More
Fisherbrand™ Replacement Deuterium Lamp for Dionex UV and Visible HPLC Detector
Replacement Deuterium Lamp for Dionex UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
$2,443.98
Specifications
Light Source Deuterium
Weight (English) 0.36 lb.
Weight (Metric) 0.16 kg
Product Type Replacement Detector Lamp
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
14500724
View Documents Promotion Details
Fisherbrand™
LDDIO105LL
UVD 3000 Nano LC D2 Detector
Each for $2,443.98
Only null left
Description
Description
Fisherbrand™ Replacement Deuterium Lamp for Dionex UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Compliance
WEEE and ROHS compliant
Specifications
Specifications
Deuterium
0.36 lb.
0.16 kg
Replacement Detector Lamp