Learn More
Fisherbrand™ Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector
Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
Supplier:  Fisherbrand™ LDAGI105LLFB
 View more versions of this product
Catalog No. 14500722
Only null left
Description
Description
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Compliance
WEEE and ROHS compliant
Specifications
Chemical Identifiers
Deuterium
0.36 lb.
0.16 kg
Detector Lamp
Specifications
Deuterium
0.16 kg
1100/1200 DAD HPLC Detector
0.36 lb.
Item 14-500-722 is comparable to Agilent part numbers 5181-1530, 2140-0813, 2140-0590.
Detector Lamp
Product Suggestions
Product Suggestions