Learn More
Eppendorf™ Repeater™ M4 Manual Handheld Pipette Dispenser PROMO
Catalog No. 14287150pm
Change view
Click to view available options
Eppendorf™ Handheld Pipette Dispenser
Catalog No.
14287150PM Eppendorf™ Handheld Pipette Dispenser
1 options
Catalog No. 14287150PM Supplier Eppendorf™ Supplier No. 2231001122
Only null left
Uses positive displacement principal to precisely and accurately dispense virtually any liquid. Dispenses liquid directly without air cushion, ensuring correct volume is dispensed regardless of density, viscosity and volatility of liquid.
  • Step counter: Displays the number of executed dispensing steps
  • Ergonomically shaped operating lever: Dispenses the liquid and ejects the Combitip advanced
  • Ergonomically shaped filling lever: For filling and emptying the Combitip advanced
  • Volume selection dial: 20 different volume settings for every Combitip size; Easy to reach and adjust using your thumb
  • Hand rest: Supports relaxed hand position and ease of use
  • Motion Sensor: Repeater switches off when not in use, and back on with minimal movements to save power and time
  • Combitip sensor: Automatic Combitip recognition and volume display
  • Embedded RFID chip
  • Supports error-free working even after an interruption or distraction during dispensing procedure
Specifications
Volume (Metric) 1 μL to 10 mL
Includes Repeater M4, Combitip Rack, Combitip Assortment Pack and Wall/Carousel Holder