Learn More
Eppendorf™ Repeater™ E3 bundle, including charger stand
Catalog No. 13683552
Change view
Click to view available options
Volume (Metric):
1 to 50 mL
Catalog No. Volume (Metric)
13683552 1 to 50 mL
1 options
Catalog No. 13683552 Supplier Eppendorf™ Supplier No. 4987000398
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The motor driven dispenser gives you full volume control, improved accuracy and reproducibility while reducing physical strain during use. The repetitive dispensing modes contribute to reducing fatigue by decreasing the number of movements for the user; filling the tip once allows dispensing up to 100 times.
  • Automatic Combitip recognition: Eliminates time consuming volume calculations, avoiding incorrect dispensing volumes
  • Ergonomic one-button tip ejector: For one handed operation and contact-free Combitips advanced ejection
  • Speed Adjustment: Program aspiration and dispensing speed for the utmost in precision and accuracy while also preventing splashing
  • Repetitive dispensing modes: Decrease the number of movements for the user by allowing them to fill the tip once and dispense up to 100 times without a refill
  • Wide dispensing range: 1 μL to 50 mL provides up to 5,000 dispensing volumes with increments as low as 100 nanoliters
  • Motor driven function reduces the human error to a minimum and reduces risk of repetitive strain injuries (RSI)
  • Safe and ergonomic operation with 9 different selectable languages
  • Large illuminated color display reduces eye strain when reading
  • Positive displacement system: Not affected by liquid properties, most manual pipetting errors and provides protection from aerosol contamination
Specifications
Number of Channels 1
Product Line Repeater E3
Type Motor Driven Dispenser
Volume (Metric) 1 to 50 mL