Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile
Catalog No. 07201488
Change view
Click to view available options
Volume (Metric):
12.5 mL
25 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
07201488 12.5 mL
07201489 25 mL
07201490 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07201488 Supplier Corning™ Supplier No. 6633
Only null left
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
  • Pyrogen-, ATP-, RNase-, and DNase-free
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO8655 ISO11137
Material Polypropylene
Sterility Sterile
For Use With (Application) Dispensing of viscous and volatile liquids
Quantity 100/Pk.
Volume (Metric) 12.5 mL