missing translation for 'onlineSavingsMsg'
Learn More

Repaglinide 98.0+%, TCI America™

Catalog No. R0179200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0179200MG 200 mg
R01791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0179200MG Supplier TCI America Supplier No. R0179200MG
Only null left

Chemical Identifiers

CAS 135062-02-1
Molecular Formula C27H36N2O4
Molecular Weight (g/mol) 452.60
MDL Number MFCD00906179
InChI Key FAEKWTJYAYMJKF-QHCPKHFHSA-N
Synonym (S)-(+)-2-Ethoxy-4-[N-[1-(2-piperidinophenyl)-3-methyl-1-butyl]aminocarbonylmethyl]benzoic Acid
PubChem CID 65981
ChEBI CHEBI:8805
IUPAC Name 2-ethoxy-4-({[(1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]carbamoyl}methyl)benzoic acid
SMILES CCOC1=CC(CC(=O)N[C@@H](CC(C)C)C2=CC=CC=C2N2CCCCC2)=CC=C1C(O)=O

Specifications

Physical Form Powder/Crystals
Color White/Yellow
Melting Point 133°C
Quantity 200 mg
Formula Weight 452.60
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Repaglinide
RTECSNumber DI0876305
TSCA No
Recommended Storage Refrigerator