missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 2

Remel PathoDx and Extraction Reagent 2 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.

Specifications

Content And Storage 2°C to 8°C
CE Marker Yes
Format Vial
Quantity Each
View More Specs

Products 1
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
R62057
View Documents Promotion Details
Thermo Scientific™
NRE4558
PathoDx Extraction Reagent 2, 14mL/Vial Each N/A
Only null left
 
Description

Description

Remel PathoDx and Extraction Reagent 2 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.

Specifications

Specifications

2°C to 8°C
Yes
Vial
Each
Videos
SDS
Documents

Documents