missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 2

Catalog No. R62057
Change view
Click to view available options
Description:
PathoDx Extraction Reagent 2, 14mL/Vial
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
R62057 PathoDx Extraction Reagent 2, 14mL/Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R62057 Supplier Thermo Scientific™ Supplier No. NRE4558

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Remel PathoDx and Extraction Reagent 2 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.

Specifications

Content And Storage 2°C to 8°C
Description PathoDx Extraction Reagent 2, 14mL/Vial
CE Marker Yes
Format Vial
Quantity Each