missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Moxalactam Selective Supplement

Catalog No. R450551
Change view
Click to view available options
Quantity:
10 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R450551 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R450551 Supplier Thermo Scientific™ Supplier No. RE450551

Remel Moxalactam Selective Supplement is used with Modified Oxford Base.

Remel Moxalactam Selective Supplement is used with Modified Oxford Base.

Specifications

Description Moxalactam Selective Supplement
Form Liquid
Product Type Selective Supplement
Quantity 10 x 10 mL
Species Moxalactam