Learn More
Reichert™ Reichert™ Rhino Ultra-Durable Handheld Refractometers
Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant
Supplier:  Reichert™ 137533L0
 View more versions of this product
Catalog No. 13-975-126
Only null left
Description
Description
Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant
  • Automatic Temperature Compensation eliminates the need for temperature correction charts
  • Optics and instrument scales are permanently sealed inside the refractometer body
  • Magnified scales have easy-to-read markings and divisions for true readings and accurate results
  • Models are offered with Brix scales of 0-30, 0-50 and 50-90%
Specifications
Specifications
Handheld Ultra-durable Refractometer
1.4201 to 1.5177
±0.2
Food and beverage, chemical, pharmaceutical and industrial cutting fluids
50 to 90 Brix
BRIX 90
0.2%
IP67
Product Suggestions
Product Suggestions