missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Regular Fetal Bovine Serum

Catalog No. MT35010CV
Change view
Click to view available options
Quantity:
500 mL
Product Type:
Fetal Bovine Serum
Heat Inactivated:
No
Yes
pH:
7.6 ±0.8
7.8 ±0.8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Heat Inactivated pH
MT35010CV 500 mL Fetal Bovine Serum No 7.6 ±0.8
MT35011CV 500 mL Fetal Bovine Serum Yes 7.8 ±0.8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MT35010CV Supplier Corning™ Supplier No. 35010CV
  • ONLINE EXCLUSIVE PRICE
    $1,009.37
    $693.64 / Each

    Save $315.73

    31.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.

  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Regular FBS is sourced from herds in USDA approved countries (ex: Mexico, Honduras, Costa Rica, Nicaragua, etc.)

Order Info

Shipped in frozen form

Specifications

Certifications/Compliance cGMP
Color Amber Yellow to Reddish
Content And Storage -25° to -15°C
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Triple-filtered, 0.1 μm
Source Bovine Fetus
Grade Regular
Endotoxin Concentration ≤20EU/ mL
For Use With (Application) Used as a supplement to cell culture media to help sustain cell growth and metabolism. This product is for research use only.
Heat Inactivated No
Hemoglobin Concentration <30 mg/dL
Osmolality 315 ± 35mOsm/kg
pH 7.6 ±0.8
Protein Content 30 to 50g/L
USDA Safety Testing Included USDA Inspected
Show More Show Less

For research use only.