missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate, Dihydrate, Technical, Spectrum™ Chemical

Catalog No. 18609541
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609541 12 kg Poly Bottle
18609539 500 g Poly Bottle
18609540 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609541 Supplier Spectrum Chemical Mfg Cor Supplier No. C116212KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10101-41-4

Specifications

Physical Form Powder, Needles or Crystals
CAS Max % 1
Quantity 12 kg
Packaging Poly Bottle
Molecular Weight (g/mol) 172.17
Grade Technical
Chemical Name or Material Calcium Sulfate, Dihydrate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.