Learn More
KOVA™ Refractrol SP Control
Tri-level control provides five levels of serum protein control solutions for use in refractometry
Supplier: KOVA™ 84679
Description
- Recovery of the assigned values provides assurance of measurements of plasma or serum protein concentration
- Each level of Refractrol is separately packaged in easy-to-use 2mL vials
Store at room temperature. KOVA Refractrol SP will remain stable in the unopened vial as specified on the product label. KOVA Refractrol SP has an open vial stability of up to 14 days.
Recommended Storage
Store at room temperature. KOVA Refractrol SP will remain stable in the unopened vial as specified on the product label. KOVA Refractrol SP has an open vial stability of up to 14 days.
Chemical Identifiers
| Store at room temperature. KOVA Refractrol SP will remain stable in the unopened vial as specified on the product label. KOVA Refractrol SP has an open vial stability of up to 14 days. | |
| Refractrol SP Control | |
| 12 x 2 mL |
| Refractrol SP Control | |
| For use with temperature compensated and non-compensated refractometers; Measurements of plasma or serum protein concentration |
Specifications
| Store at room temperature. KOVA Refractrol SP will remain stable in the unopened vial as specified on the product label. KOVA Refractrol SP has an open vial stability of up to 14 days. | |
| Refractrol SP Control | |
| 6 |
| Refractrol SP Control | |
| For use with temperature compensated and non-compensated refractometers; Measurements of plasma or serum protein concentration | |
| 12 x 2 mL |