Learn More
KOVA™ Refractrol SP Control
Catalog No. p-4839997
Change view
Click to view available options
Level:
6
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
22749770 9
22749769 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22749770 Supplier KOVA™ Supplier No. K84655
Only null left
Tri-level control provides five levels of serum protein control solutions for use in refractometry
  • Recovery of the assigned values provides assurance of measurements of plasma or serum protein concentration
  • Each level of Refractrol is separately packaged in easy-to-use 2mL vials
Storage:
Store at room temperature. KOVA Refractrol SP will remain stable in the unopened vial as specified on the product label. KOVA Refractrol SP has an open vial stability of up to 14 days.
Recommended Storage
Store at room temperature. KOVA Refractrol SP will remain stable in the unopened vial as specified on the product label. KOVA Refractrol SP has an open vial stability of up to 14 days.
Specifications
Content And Storage Store at room temperature. KOVA Refractrol SP will remain stable in the unopened vial as specified on the product label. KOVA Refractrol SP has an open vial stability of up to 14 days.
Description Refractrol SP Control
Type Refractrol SP Control
Intended Use For use with temperature compensated and non-compensated refractometers; Measurements of plasma or serum protein concentration
Level 9
Quantity 12 x 2 mL