Learn More
MilliporeSigma™ Reflectoquant™ Peroxide Test Strips
For determination of inorganic peroxides in aqueous solutions
Supplier: MilliporeSigma™ 1.16731.0001
Includes: Tube containing 50 strips, 1 bar-code strip.
Description
- Enzymatic reaction method ideal for screening
- Barcode calibration for reliable, quantitative results
- Low analysis costs
Reclose the tube containing the test strips immediately after use.
For food and water analysis
Chemical Identifiers
| For food and water analysis | |
| 2°C to 8°C |
Specifications
| For food and water analysis | |
| 100 to 1000mg/L | |
| 100 to 1000mg/L | |
| Enzymatic Reaction |
| Reflectometer | |
| 50 | |
| 2°C to 8°C | |
| Test Strip |