missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NoTrax™ Reflective Parking Stop Curb Park-It™

Park-It™ car stops made of 100% recycled rubber, used in parking lots to effectively stop cars pulling into parking spots
Supplier: NoTrax™ GNRS1610YB
Description
- Constructed of 100% recycled rubber
- Used in parking lots to stop cars pulling into parking spots
- More forgiving of vehicles, sturdier, and do not break or need to be replaced
- In-molded, highly reflective tape in yellow or white, to ensure visibility and pedestrian safety
- Recycled rubber stops pre-drilled with holes
- Flexible rubber design conforms to contour of uneven road surfaces
- ADA Handicap Park-It™ is designed specifically for handicap parking spaces
- Blue and white contrasting colors are immediately identifiable to differentiate handicap parking spaces to vehicles and pedestrians
Chemical Identifiers
| Recycled Rubber |
Specifications
| Recycled Rubber | |
| 6 in. | |
| Parking Stop | |
| Black and Yellow Striped |