missing translation for 'onlineSavingsMsg'
Learn More

EP Reference Solution Y4, Yellow Reference Solution, Ricca Chemical

Catalog No. 288344
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
288344 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 288344 Supplier Ricca Chemical Company Supplier No. 2883.44

Specifications

Chemical Name or Material Reference solution Y4
Packaging Amber Glass Bottle
CAS 10025-77-1, 7647-01-0, 7732-18-5, 7758-99-8, 7791-13-1
Color Yellow
pH Acidic
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Yellow standard solution
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 120 mL
Grade EP
Show More Show Less