missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Reference™ 2, Mechanical Multi-Channel Pipette
GREENER_CHOICE

Catalog No. 05412451
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 100 μL
30 to 300 μL
Number of Channels:
12
8
Color:
Medium Gray
Orange
Yellow
Increments:
0.01 μL
0.1 μL
0.2 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels Color Increments
05412451 30 to 300 μL 8 Orange 0.2 μL
05412449 0.5 to 10 μL 8 Medium Gray 0.01 μL
05412450 10 to 100 μL 8 Yellow 0.1 μL
05412452 0.5 to 10 μL 12 Medium Gray 0.01 μL
05412453 10 to 100 μL 12 Yellow 0.1 μL
05412454 30 to 300 μL 12 Orange 0.2 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 05412451 Supplier Eppendorf™ Supplier No. 4926000050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Minimize risk of aerosol contamination and reduce thumb strain with these variable volume pipettes. Eppendorf™ Reference™ 2 Variable Volume Pipettes are ideal for pipetting and mixing liquids, filling of plates, gels and reaction vessels, and phase extraction and removal of supernatants.

  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Single-button operation
  • Has Spring-loaded tip cone, movable parts, stainless steel external edges with customizable labeling area
  • Ergonomic handling with reduced operating effort
  • Simple and quick tip ejection with active aerosol reduction
  • Reduce repetitive strain injuries by not having to move your thumb sideways
  • Haptic feedback before tip ejection
  • High precision and accuracy provides reliable pipetting results
  • User friendly secondary adjustment

Specifications

Autoclavable Autoclavable
Color Orange
For Use With Pipette Tip 300 μL
Number of Channels 8
Product Line Reference 2
Type Variable Volume Pipette
Volume (Metric) 30 to 300 μL
For Use With (Equipment) For use with 300 μL pipette tips
Includes epT.I.P.S.™ Box 2.0 with 96 Tips
Increments 0.2 μL