missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Reference™ 2, Mechanical Multi-Channel Pipette

Minimize risk of aerosol contamination and reduce thumb strain with these variable volume pipettes. Eppendorf™ Reference™ 2 Variable Volume Pipettes are ideal for pipetting and mixing liquids, filling of plates, gels and reaction vessels, and phase extraction and removal of supernatants.

Supplier:  Eppendorf™ 4926000034

 View more versions of this product

Catalog No. 05-412-450


Only null left

Description

Description

  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust
  • High flexibility with ability to remove single cones
  • Ensures continuous identical pipet alignment during work process
  • Fully autoclavable pipet with smooth surface and few rough edges
  • Single-button operation
  • Has Spring-loaded tip cone, movable parts, stainless steel external edges with customizable labeling area
  • Ergonomic handling with reduced operating effort
  • Simple and quick tip ejection with active aerosol reduction
  • Reduce repetitive strain injuries by not having to move your thumb sideways
  • Haptic feedback before tip ejection
  • High precision and accuracy provides reliable pipetting results
  • User friendly secondary adjustment
Specifications

Specifications

Autoclavable
Pipette Tip 200 μL
Reference 2
10 to 100 μL
epT.I.P.S.™ Box 2.0 with 96 Tips
Yellow
8
Variable Volume Pipette
For use with 200 μL pipette tips
0.1 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions