Learn More
United Scientific™ Reducing Adapter
Effortlessly connect glassware of varying sizes with this Reducing Adapter, featuring a smaller socket joint at the top and a larger socket joint at the bottom for seamless integration.
Supplier:  United Scientific™ UNRDSOK2445
 View more versions of this product
Catalog No. S012419
Only null left
Explore available promotions
Description
Description
  • Made of borosilicate glass 3.3
  • Chemical-resistant
  • ASTM E 438 Type 1 Class A
  • Meets DIN/ISO 3585 and USP Type I requirement
  • Autoclavable
Specifications
Specifications
45/50
Borosilicate Glass (3.3)
Reducing Adapter
24/40
Product Suggestions
Product Suggestions