missing translation for 'onlineSavingsMsg'
Learn More

Redox Standard Solution, Iodide/Triiodide, Ricca Chemical

Numéro de catalogue. 659532
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
659532 1 L
659516 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 659532 Fournisseur Ricca Chemical Company Code fournisseur 659532
Il en reste null

Spécifications

Chemical Name or Material Redox Standard Solution
Packaging Natural Poly Bottle
Name Note Iodide/Triiodide
CAS 7681-11-0, 7553-56-2, 10043-35-3, 1310-58-3, 7732-18-5
CAS Min % 59.0, 38.31, 0.73, 0.1, 0.07
CAS Max % 61.0, 39.87, 0.76, 0.11, 0.08
Physical Form Liquid
Color Brown
Quantity 1 L