missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Redox Standard Solution, Iodide/Triiodide, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Redox Standard Solution |
| Packaging | Natural Poly Bottle |
| Name Note | Iodide/Triiodide |
| CAS | 7681-11-0, 7553-56-2, 10043-35-3, 1310-58-3, 7732-18-5 |
| CAS Min % | 59.0, 38.31, 0.73, 0.1, 0.07 |
| CAS Max % | 61.0, 39.87, 0.76, 0.11, 0.08 |
| Physical Form | Liquid |
| Color | Brown |
| Quantity | 500 mL |