missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-it™ RiboGreen Reagent and RNA Assay Kit

Catalog No. R32700
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 mL
Product Type:
Quant-iT RiboGreen RNA Kit
Quant-iT RiboGreen RNA Reagent
RediPlate 96 RiboGreen RNA Kit
Quantitation Range:
1 to 200 ng
3 to 200 ng
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Quantitation Range
R32700 1 kit RediPlate 96 RiboGreen RNA Kit 3 to 200 ng
R11490 1 kit Quant-iT RiboGreen RNA Kit 1 to 200 ng
R11491 1 mL Quant-iT RiboGreen RNA Reagent 1 to 200 ng
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R32700 Supplier Invitrogen™ Supplier No. R32700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

20X TE buffer, ribosomal RNA standards, one 96-well microplate with 12 removable strips, each with 8 wells. Eleven strips are pre-loaded with RiboGreen Reagent. Remaining strip (blackened tabs) contains a series of RNA for generating standard curves.

Quant-iT RiboGreen RNA Reagent is an ultrasensitive fluorescent nucleic acid stain for quantitating RNA in solution. The Quant-iT RiboGreen RNA Assay Kit comes with reagent, dilution buffer, and rRNA standards to provide a linear detection range of 1–200 ng of RNA.

Quant-iT RiboGreen RNA Assay Kit
Containing one of the most sensitive detection dyes for the quantitation of RNA, the Quant-iT RiboGreen RNA Assay Kit and the RediPlate 96 RiboGreen RNA Quantitation Kit offer linear fluorescence detection in the range of 1–200 ng of RNA. Both assay kits contain Quant-iT RiboGreen RNA reagent as well as buffers and RNA standards.

RediPlate 96 RiboGreen RNA Quantitation Kit
The RediPlate 96 RiboGreen RNA Quantitation Kit is a rapid, simple, and highly sensitive fluorescence-based RNA-quantitation assay. The microplate is pre-loaded with RiboGreen reagent, which shows bright green fluorescence upon binding to RNA or DNA. Just add buffer and samples to the microplate wells, wait 10 minutes, and read the fluorescence in any standard fluorescence-based microplate reader. The versatile RediPlate 96 design is ideal for either high-throughput RNA quantitation or the measurement of a small sample numbers.

The RediPlate 96 RiboGreen RNA Quantitation Kit also offers the following benefits:
• Automatable—a 10 minute incubation time with no separation steps makes the assay ideal for automated, high-throughput measurements
• Precise—the average coefficient of variation for replicate assays is ≤5% using a robotic liquid handling system
• Convenient—each plate includes pre-measured RNA standards and is ready for immediate use
• Easy to read—the assay can be performed using standard fluorescence-based microplate readers

Applications
The RediPlate 96 RiboGreen RNA Quantitation Kit is ideal for measuring yields of RNA preparations for microarray experiments, reverse transcription PCR (RT-PCR), differential display PCR, northern blot analysis, S1 nuclease assays, RNase protection assays, and cDNA library preparations.

Quant-iT RiboGreen RNA Reagent
The Quant-iT RiboGreen RNA Reagent is one of the most sensitive fluorescent dyes for the quantitation of RNA in solution, with linear fluorescence detection in the range of 1-200 ng of RNA. This reagent is also included with the Quant-iT RiboGreen RNA Assay Kit and comes complete with buffer and RNA standard.

Order Info

Shipping Condition: Room temperature

Specifications

Detection Method Fluorescence
Excitation/Emission 500/525
For Use With (Equipment) Microplate Reader
Quantitation Range 3 to 200 ng
Product Line RIBOGREEN, RediPlate
Product Type RediPlate 96 RiboGreen RNA Kit
Quantity 1 kit
Shipping Condition Room Temperature
When I try to run RediPlate 96 RiboGreen RNA Kit (Cat. No. R32700) on my Varioskan LUX instrument, it says there is no plate. What should I do?

When using a strip plate with the Varioskan LUX plate reader, ensure that there is a strip covering the D6 well, as this is where the instrument checks for the presence of a plate. The strip does not need to be active; it just needs to be in place for the instrument to verify the plate's presence. Alternatively, you can resolve this issue by adjusting the software settings. Go to the general settings and uncheck the box labeled "Check plate and tip prime vessel before session execution." This will immediately solve the problem.

Does the Quant-it RiboGreen RNA Assay Kit and RiboGreen RNA Reagent (Cat. No. R32700) work with double-stranded RNA (dsRNA)?

We have not validated measurement of dsRNA with the Quant-it RiboGreen RNA Assay Kit and RiboGreen RNA Reagent (Cat. No. R32700). If you are using RiboGreen dye to measure dsRNA, then you should use your own dsRNA standards, rather than the standards provided with the kit, to get an accurate measurement.