missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Red PTFE/Silicone Septa

Vial septa are available in a variety of materials

Supplier:  MilliporeSigma™ Supelco™ 29039U

 View more versions of this product

Catalog No. 111012013


Only null left

Description

Description

  • Vial septa are available in a variety of materials.
  • Septa, tan PTFE/silicone are sized to fit screw top vials.
  • They are suitable for use with most organics (until pierced) and are not recommended for use with chlorosilanes.
Specifications

Chemical Identifiers

PTFE/Silicone
Temperature limit 120°C

Specifications

9 mm
PTFE/Silicone
Temperature limit 120°C
Red
100
1 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions