missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Minigrip™ Red Line™ Plain Double Zipper Bags
Leak-resistant bags which fits any industrial need. Minigrip Red Line Plain Double Zipper Bags feature easy open/close zipper and manufactured with 100% FDA-approved virgin LDPE and LLDPE resins.
Supplier: Minigrip™ MGRL4P1424
Description
- Premium zipper easily snaps open
- Manufactured with 100% FDA-approved virgin LDPE and LLDPE resins
- Reinforced zipper and side seams to keep bags from splitting when opened or filled
Specifications
| Double Zipper | |
| LDPE, LLDPE | |
| 35.5 x 60.9cm (14 x 24 in.) | |
| Sample Bag |