missing translation for 'onlineSavingsMsg'
Learn More

Red Kap™ Twill Action Back Coveralls

Coveralls give flexibility to work with ample range of motion for reaching, squatting, and more. Oversized for a roomy fit and can be worn over clothes.

Supplier:  Red Kap™ CT10ORRG44

 View more versions of this product

Catalog No. 19-295-007


Only null left
Explore available promotions

Description

Description

  • 7.25 oz. twill fabric
  • 65% polyester/35% combed cotton blend
  • Care: home or industrial wash
  • Two-way brass zipper with gripper at top and at lapel
  • One-piece topstitched collar
  • Two set-in front pockets, two patch hip pockets, two chest pockets, rule leg pocket
  • Four-needle band joins top and bottom
  • Safety stitched outseams, felled inseams
  • Hemmed sleeves; action back; side vent opening
  • Sized to be worn over clothes
Specifications

Specifications

General Purpose Coveralls
Men's
Orange
2-way Zipper
Topstitched
Yes
44 in.
111.75 cm
Hemmed
Regular
44
Polyester, Combed Cotton
Open
Non-sterile
7.25 oz. Twill Construction, Finish: Pre-cure Durable Press with Soil Release
7
44 in.
111.75 cm
Manufacturing, Maintenance, Automotive
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions